6-Methyl-2-mercaptobenzothiazole 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methylbenzo[d]thiazole-2-thiol

OR958424-1G
APOSOR958424-1GEA 56.7 EUR
APOSOR958424-1G
6-Methyl-2-mercaptobenzothiazole 95%
6-Methyl-2-mercaptobenzothiazole
Formel: C₈H₇NS₂
Molecular Weight: 181.278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774732
CAS-Nummer: 2268-79-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR