(R)-b-Amino-4-methoxy-benzeneethanol 97%

Lieferant: Apollo Scientific

OR958581-1G OR958581-250MG OR958581-500MG
APOSOR958581-1GEA 783 EUR
APOSOR958581-1G APOSOR958581-250MG APOSOR958581-500MG
(R)-b-Amino-4-methoxy-benzeneethanol 97%
(R)-b-Amino-4-methoxy-benzeneethanol
Formel: C₉H₁₃NO₂
Molecular Weight: 167,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09253729
CAS-Nummer: 100929-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR