Ethyl-2-[[(2'-cyanobiphenyl-4-yl)methyl]amino]-3-nitrobenzoate 95%

Lieferant: Apollo Scientific

OR958836-100MG OR958836-1G OR958836-250MG
APOSOR958836-100MGEA 126 EUR
APOSOR958836-100MG APOSOR958836-1G APOSOR958836-250MG
Ethyl-2-[[(2'-cyanobiphenyl-4-yl)methyl]amino]-3-nitrobenzoate 95%
Ethyl-2-[[(2'-cyanobiphenyl-4-yl)methyl]amino]-3-nitrobenzoate
Formel: C₂₃H₁₉N₃O₄
Molecular Weight: 401,415 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD09030634
CAS-Nummer: 136285-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR