2-Methyl-1-phenylbutane-1,3-dione 95%
Lieferant: Apollo Scientific
OR959238-1G
OR959238-250MG
APOSOR959238-1GEA
98.6
EUR
APOSOR959238-1G
APOSOR959238-250MG
2-Methyl-1-phenylbutane-1,3-dione 95%
2-Methyl-1-phenylbutane-1,3-dione
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033549 CAS-Nummer: 6668-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...