(R)-Piperazin-2-ylmethanol dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR959303-100MG OR959303-250MG
APOSOR959303-100MGEA 33.5 EUR
APOSOR959303-100MG APOSOR959303-250MG
(R)-Piperazin-2-ylmethanol dihydrochloride 97%
(R)-Piperazin-2-ylmethanol dihydrochloride
Formel: C₅H₁₂N₂O·2ClH
Molecular Weight: 189,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954306
CAS-Nummer: 1217482-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR