3,5-dichloro-4-methylpyridin-2-ol 95%

Lieferant: Apollo Scientific

OR959396-1G OR959396-250MG
APOSOR959396-1GEA 348 EUR
APOSOR959396-1G APOSOR959396-250MG
3,5-dichloro-4-methylpyridin-2-ol 95%
3,5-dichloro-4-methylpyridin-2-ol
Formel: C₆H₅Cl₂NO
Molecular Weight: 178.016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16883065
CAS-Nummer: 58236-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR