2,3,5-Trichloro-4-methylpyridine 98%
Lieferant: Apollo Scientific
OR959397-1G
OR959397-25G
OR959397-5G
APOSOR959397-1GEA
145
EUR
APOSOR959397-1G
APOSOR959397-25G
APOSOR959397-5G
2,3,5-Trichloro-4-methylpyridine 98%
2,3,5-Trichloro-4-methylpyridine
Formel:
C₆H₄Cl₃N Molecular Weight: 196,462 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20529523 CAS-Nummer: 53939-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...