Methyl 2-amino-5-chloropyridine-4-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-amino-5-chloroisonicotinate
OR959406-1G
OR959406-5G
APOSOR959406-1GEA
94.4
EUR
APOSOR959406-1G
APOSOR959406-5G
Methyl 2-amino-5-chloropyridine-4-carboxylate 98%
Methyl 2-amino-5-chloropyridine-4-carboxylate
Formel:
C₇H₇ClN₂O₂ Molecular Weight: 186.596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22375099 CAS-Nummer: 1227002-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...