2-Methoxy-5-phenylpyridine 95%

Lieferant: Apollo Scientific

OR959445-1G OR959445-250MG
APOSOR959445-1GEA 852 EUR
APOSOR959445-1G APOSOR959445-250MG
2-Methoxy-5-phenylpyridine 95%
2-Methoxy-5-phenylpyridine
Formel: C₁₂H₁₁NO
Molecular Weight: 185,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044373
CAS-Nummer: 53698-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR