7,8-Dimethylquinoline 98%
Lieferant: Apollo Scientific
OR959482-1G
OR959482-5G
APOSOR959482-1GEA
270
EUR
APOSOR959482-1G
APOSOR959482-5G
7,8-Dimethylquinoline 98%
7,8-Dimethylquinoline
Formel:
C₁₁H₁₁N Molecular Weight: 157,212 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18448797 CAS-Nummer: 20668-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...