6-(Isopropylamino)pyridine-3-carbonitrile 98%

Lieferant: Apollo Scientific

OR959604-10G OR959604-1G OR959604-5G
APOSOR959604-10GEA 754 EUR
APOSOR959604-10G APOSOR959604-1G APOSOR959604-5G
6-(Isopropylamino)pyridine-3-carbonitrile 98%
6-(Isopropylamino)pyridine-3-carbonitrile
Formel: C₉H₁₁N₃
Molecular Weight: 161,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09944214
CAS-Nummer: 160017-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR