methyl 5-methoxypyridine-2-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-methoxypicolinate

OR959687-5G
APOSOR959687-5GEA 200 EUR
APOSOR959687-5G
methyl 5-methoxypyridine-2-carboxylate 98%
methyl 5-methoxypyridine-2-carboxylate
Formel: C₈H₉NO₃
Molecular Weight: 167,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11975090
CAS-Nummer: 29681-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR