3-Ethoxypyrazine-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR959696-1G OR959696-5G
APOSOR959696-1GEA 219 EUR
APOSOR959696-1G APOSOR959696-5G
3-Ethoxypyrazine-2-carboxylic acid 98%
3-Ethoxypyrazine-2-carboxylic acid
Formel: C₇H₈N₂O₃
Molecular Weight: 168.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16852854
CAS-Nummer: 1339121-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR