5-Chloro-3-methoxypyridin-2-ol 98%

Lieferant: Apollo Scientific

OR959700-1G OR959700-5G
APOSOR959700-1GEA 161 EUR
APOSOR959700-1G APOSOR959700-5G
5-Chloro-3-methoxypyridin-2-ol 98%
5-Chloro-3-methoxypyridin-2-ol
Formel: C₆H₆ClNO₂
Molecular Weight: 159.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415703
CAS-Nummer: 94135-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR