6-Chloro-5-methoxypyridine-3-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-5-methoxynicotinaldehyde

OR959701-1G OR959701-250MG
APOSOR959701-1GEA 806 EUR
APOSOR959701-1G APOSOR959701-250MG
6-Chloro-5-methoxypyridine-3-carbaldehyde 95%
6-Chloro-5-methoxypyridine-3-carbaldehyde
Formel: C₇H₆ClNO₂
Molecular Weight: 171,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189003
CAS-Nummer: 1060801-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR