Methyl 4-chloro-6-methylpyridine-3-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-chloro-6-methylnicotinate

OR959791-1G OR959791-250MG
APOSOR959791-1GEA 53.8 EUR
APOSOR959791-1G APOSOR959791-250MG
Methyl 4-chloro-6-methylpyridine-3-carboxylate 97%
Methyl 4-chloro-6-methylpyridine-3-carboxylate
Formel: C₈H₈ClNO₂
Molecular Weight: 185.608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374919
CAS-Nummer: 886372-05-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR