N-Methoxy-N-methylpentanamide 95%

Lieferant: Apollo Scientific

OR959866-1G OR959866-5G
APOSOR959866-1GEA 81.2 EUR
APOSOR959866-1G APOSOR959866-5G
N-Methoxy-N-methylpentanamide 95%
N-Methoxy-N-methylpentanamide
Formel: C₇H₁₅NO₂
Molecular Weight: 145.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05149014
CAS-Nummer: 129118-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR