1-Butylpyrazole 98%

Lieferant: Apollo Scientific

Synonyme: 1-Butyl-1H-pyrazole

OR959882-1G OR959882-5G
APOSOR959882-1GEA 74.1 EUR
APOSOR959882-1G APOSOR959882-5G
1-Butylpyrazole 98%
1-Butylpyrazole
Formel: C₇H₁₂N₂
Molecular Weight: 124,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08701137
CAS-Nummer: 52096-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR