2-(Cyclopropylamino)benzonitrile 97%

Lieferant: Apollo Scientific

OR959901-10G OR959901-1G OR959901-5G
APOSOR959901-10GEA 754 EUR
APOSOR959901-10G APOSOR959901-1G APOSOR959901-5G
2-(Cyclopropylamino)benzonitrile 97%
2-(Cyclopropylamino)benzonitrile
Formel: C₁₀H₁₀N₂
Molecular Weight: 158.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11155689
CAS-Nummer: 675575-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR