2-(Ethylamino)benzonitrile 98%
Lieferant: Apollo Scientific
OR959906-1G
OR959906-5G
APOSOR959906-1GEA
145
EUR
APOSOR959906-1G
APOSOR959906-5G
2-(Ethylamino)benzonitrile 98%
2-(Ethylamino)benzonitrile
Formel:
C₉H₁₀N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11144683 CAS-Nummer: 30091-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...