2-(Propylamino)benzonitrile 98%
Lieferant: Apollo Scientific
OR959910-10G
OR959910-1G
OR959910-5G
APOSOR959910-10GEA
538
EUR
APOSOR959910-10G
APOSOR959910-1G
APOSOR959910-5G
2-(Propylamino)benzonitrile 98%
2-(Propylamino)benzonitrile
Formel:
C₁₀H₁₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11155673 CAS-Nummer: 74396-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...