1-Cyclohexylpyrazole 95%
Lieferant: Apollo Scientific
OR960026-10G
OR960026-1G
OR960026-5G
APOSOR960026-10GEA
754
EUR
APOSOR960026-10G
APOSOR960026-1G
APOSOR960026-5G
1-Cyclohexylpyrazole 95%
1-Cyclohexylpyrazole
Formel:
C₉H₁₄N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD14609485 CAS-Nummer: 97421-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...