2-Chloro-N-methoxy-N-methylbenzamide 97%

Lieferant: Apollo Scientific

OR960086-1G OR960086-5G
APOSOR960086-1GEA 163 EUR
APOSOR960086-1G APOSOR960086-5G
2-Chloro-N-methoxy-N-methylbenzamide 97%
2-Chloro-N-methoxy-N-methylbenzamide
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684306
CAS-Nummer: 289686-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR