4-(Thiophen-2-ylmethyl)morpholine 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Thiophen-2-ylmethyl)mopholine

OR960178-1G OR960178-5G
APOSOR960178-1GEA 219 EUR
APOSOR960178-1G APOSOR960178-5G
4-(Thiophen-2-ylmethyl)morpholine 98%
4-(Thiophen-2-ylmethyl)morpholine
Formel: C₉H₁₃NOS
Molecular Weight: 183,271 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01309097
CAS-Nummer: 338454-48-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR