2,6-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine 98%

Lieferant: Apollo Scientific

OR960208-1G OR960208-5G
APOSOR960208-1GEA 219 EUR
APOSOR960208-1G APOSOR960208-5G
2,6-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine 98%
2,6-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11603425
CAS-Nummer: 58959-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR