7-Methyl-3,4-dihydro-2H-1,4-benzoxazine 98%

Lieferant: Apollo Scientific

Synonyme: 7-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine

OR960213-1G OR960213-250MG
APOSOR960213-1GEA 121 EUR
APOSOR960213-1G APOSOR960213-250MG
7-Methyl-3,4-dihydro-2H-1,4-benzoxazine 98%
7-Methyl-3,4-dihydro-2H-1,4-benzoxazine
Formel: C₉H₁₁NO
Molecular Weight: 149.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17014030
CAS-Nummer: 71472-58-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR