1,7-Dichloroisoquinoline 97%

Lieferant: Apollo Scientific

OR960813-1G OR960813-5G
APOSOR960813-1GEA 89.9 EUR
APOSOR960813-1G APOSOR960813-5G
1,7-Dichloroisoquinoline 97%
1,7-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
Molecular Weight: 198.049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846494
CAS-Nummer: 70810-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR