Dihydro cuminyl alcohol 95%

Lieferant: Apollo Scientific

Synonyme: Perillyl alcohol, (4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl)methanol

OR961069-100G
APOSOR961069-100GEA 186 EUR
APOSOR961069-100G
Dihydro cuminyl alcohol 95%
Dihydro cuminyl alcohol
Formel: C₁₀H₁₆O
Molecular Weight: 152,233 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001567
CAS-Nummer: 536-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR