FMOC-Hmd HCl

Lieferant: Apollo Scientific

Synonyme: (9H-Fluoren-9-yl)methyl (6-aminohexyl)carbamate hydrochloride

OR961116-1G OR961116-250MG OR961116-5G
APOSOR961116-1GEA 120 EUR
APOSOR961116-1G APOSOR961116-250MG APOSOR961116-5G
FMOC-Hmd HCl
FMOC-Hmd HCl
Formel: C₂₁H₂₆N₂O₂·ClH
Molecular Weight: 374,904 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00830743
CAS-Nummer: 945923-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR