(R)-3-Amino-3-phenylpropionic acid 97%

Lieferant: Apollo Scientific

Synonyme: H-D-β-Phenyl-Ala-OH, 3-Amino-3-phenylpropionic-acid, (R)-3-Amino-3-phenylpropanoic acid, (R)-b-Amino-benzenepropanoic acid

OR961148-5G
APOSOR961148-5GEA 128 EUR
APOSOR961148-5G
(R)-3-Amino-3-phenylpropionic acid 97%
(R)-3-Amino-3-phenylpropionic acid
Formel: C₉H₁₁NO₂
Molecular Weight: 165,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01076237
CAS-Nummer: 13921-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR