Fmoc-L-4-carbamoylphe 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-carbamoylphenyl)propanoic acid
OR961170-1G
OR961170-250MG
OR961170-5G
APOSOR961170-1GEA
151
EUR
APOSOR961170-1G
APOSOR961170-250MG
APOSOR961170-5G
Fmoc-L-4-carbamoylphe 97%
Fmoc-L-4-carbamoylphe
Formel:
C₂₅H₂₂N₂O₅ Molecular Weight: 430,453 g/mol Schmelzpunkt: 243…249 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01317021 CAS-Nummer: 204716-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...