Benzyl-N-methyl-L-alanine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-2-[Benzyl(methyl)amino]propanoic acid

OR961236-1G OR961236-5G
APOSOR961236-1GEA 154 EUR
APOSOR961236-1G APOSOR961236-5G
Benzyl-N-methyl-L-alanine 96%
Benzyl-N-methyl-L-alanine
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01632093
CAS-Nummer: 63238-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR