(1-Boc-aminocyclohexyl)acetic acid 97%

Lieferant: Apollo Scientific

OR961369-1G OR961369-500MG OR961369-5G
APOSOR961369-1GEA 340 EUR
APOSOR961369-1G APOSOR961369-500MG APOSOR961369-5G
(1-Boc-aminocyclohexyl)acetic acid 97%
(1-Boc-aminocyclohexyl)acetic acid
Formel: C₁₃H₂₃NO₄
Molecular Weight: 257,326 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02258133
CAS-Nummer: 187610-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR