2-(Tetrahydro-2H-pyran-2-yl)ethanamine hydrochloride 95%

Lieferant: Apollo Scientific

OR961477-1G OR961477-250MG
APOSOR961477-1GEA 864 EUR
APOSOR961477-1G APOSOR961477-250MG
2-(Tetrahydro-2H-pyran-2-yl)ethanamine hydrochloride 95%
2-(Tetrahydro-2H-pyran-2-yl)ethanamine hydrochloride
Formel: C₇H₁₅NO·ClH
Molecular Weight: 165,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06739026
CAS-Nummer: 1005756-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR