1-Amino-cyclobutane-carboxylic acid ethyl ester hydrochloride 96%
Lieferant: Apollo Scientific
Synonyme: Ethyl 1-aminocyclobutanecarboxylate hydrochloride
OR961594-1G OR961594-5G
APOSOR961594-1GEA 82.8 EUR
APOSOR961594-1G APOSOR961594-5G
1-Amino-cyclobutane-carboxylic acid ethyl ester hydrochloride 96%
1-Amino-cyclobutane-carboxylic acid ethyl ester hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664344
CAS-Nummer: 145143-60-8