1-Benzyl-pyrrolidine 98%

Lieferant: Apollo Scientific

Synonyme: 1-Benzylpyrrolidine

OR961673-5G
APOSOR961673-5GEA 63.8 EUR
APOSOR961673-5G
1-Benzyl-pyrrolidine 98%
1-Benzyl-pyrrolidine
Formel: C₁₁H₁₅N
Molecular Weight: 161,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127807
CAS-Nummer: 29897-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR