(1-Benzylazetidin-2-yl)methanamine 95%

Lieferant: Apollo Scientific

OR961750-1G OR961750-250MG
APOSOR961750-1GEA 806 EUR
APOSOR961750-1G APOSOR961750-250MG
(1-Benzylazetidin-2-yl)methanamine 95%
(1-Benzylazetidin-2-yl)methanamine
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658338
CAS-Nummer: 46193-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR