(S)-b-Amino-4-methyl-benzeneethanol 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-2-(p-tolyl)ethanol
OR961799-1G
OR961799-250MG
APOSOR961799-1GEA
409
EUR
APOSOR961799-1G
APOSOR961799-250MG
(S)-b-Amino-4-methyl-benzeneethanol 97%
(S)-b-Amino-4-methyl-benzeneethanol
Formel:
C₉H₁₃NO Molecular Weight: 151,206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09253778 CAS-Nummer: 327183-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...