1-Methyl-1-pyridin-3-yl-ethylamine 95%
Lieferant: Apollo Scientific
OR961973-100MG
OR961973-1G
OR961973-250MG
APOSOR961973-100MGEA
89.9
EUR
APOSOR961973-100MG
APOSOR961973-1G
APOSOR961973-250MG
1-Methyl-1-pyridin-3-yl-ethylamine 95%
1-Methyl-1-pyridin-3-yl-ethylamine
Formel:
C₈H₁₂N₂ Molecular Weight: 136.194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08729280 CAS-Nummer: 99980-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...