N-Fmoc-isoindoline-1-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(((9H-Fluoren-9-yl)methoxy)carbonyl)isoindoline-1-carboxylic acid

OR961993-1G OR961993-250MG
APOSOR961993-1GEA 381 EUR
APOSOR961993-1G APOSOR961993-250MG
N-Fmoc-isoindoline-1-carboxylic acid 97%
N-Fmoc-isoindoline-1-carboxylic acid
Formel: C₂₄H₁₉NO₄
Molecular Weight: 385,412 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00270204
CAS-Nummer: 204320-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR