2-(Pyridin-4-yl)quinazolin-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxy-2-(pyridin-4-yl)quinazoline, 4-(4-Hydroxyquinazolin-2-yl)pyridine
OR9622-1G
OR9622-5G
APOSOR9622-1GEA
139
EUR
APOSOR9622-1G
APOSOR9622-5G
2-(Pyridin-4-yl)quinazolin-4-ol
2-(Pyridin-4-yl)quinazolin-4-ol
Formel:
C₁₃H₉N₃O Molecular Weight: 223,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665826 CAS-Nummer: 6484-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...