Fmoc-Ala(β-cyclobutyl)-OH 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclobutylpropanoic acid, Fmoc-β-Cyclobutyl-Ala-OH

OR962218-1G
APOSOR962218-1GEA 75.4 EUR
APOSOR962218-1G
Fmoc-Ala(β-cyclobutyl)-OH 97%
Fmoc-Ala(β-cyclobutyl)-OH
Formel: C₂₂H₂₃NO₄
Molecular Weight: 365,422 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840400
CAS-Nummer: 478183-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR