Methyl 3-(2-aminophenyl)acrylate 97%

Lieferant: Apollo Scientific

OR962459-1G OR962459-5G
APOSOR962459-1GEA 122 EUR
APOSOR962459-1G APOSOR962459-5G
Methyl 3-(2-aminophenyl)acrylate 97%
Methyl 3-(2-aminophenyl)acrylate
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00031415
CAS-Nummer: 1664-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR