Methyl 3-(4-cyanophenyl)acrylate 98%

Lieferant: Apollo Scientific

OR962483-25G OR962483-5G
APOSOR962483-25GEA 491 EUR
APOSOR962483-25G APOSOR962483-5G
Methyl 3-(4-cyanophenyl)acrylate 98%
Methyl 3-(4-cyanophenyl)acrylate
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00157182
CAS-Nummer: 67472-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR