1-(3,4-Dichlorophenyl)cyclopropanecarboxylic acid 97%
Lieferant: Apollo Scientific
OR962512-1G
APOSOR962512-1GEA
97.3
EUR
APOSOR962512-1G
1-(3,4-Dichlorophenyl)cyclopropanecarboxylic acid 97%
1-(3,4-Dichlorophenyl)cyclopropanecarboxylic acid
Formel:
C₁₀H₈Cl₂O₂ Molecular Weight: 231,075 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00461541 CAS-Nummer: 342386-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...