2-Methyl-4-quinolinecarbonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
2-Methylquinoline-4-carbonitrile
OR962547-100MG
OR962547-1G
OR962547-250MG
APOSOR962547-100MGEA
133
EUR
APOSOR962547-100MG
APOSOR962547-1G
APOSOR962547-250MG
2-Methyl-4-quinolinecarbonitrile 98%
2-Methyl-4-quinolinecarbonitrile
Formel:
C₁₁H₈N₂ Molecular Weight: 168,195 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01085095 CAS-Nummer: 29196-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...