N-Methyl-4-nitrobenzamide 98%

Lieferant: Apollo Scientific

OR962552-1G OR962552-5G
APOSOR962552-1GEA 154 EUR
APOSOR962552-1G APOSOR962552-5G
N-Methyl-4-nitrobenzamide 98%
N-Methyl-4-nitrobenzamide
Formel: C₈H₈N₂O₃
Molecular Weight: 180,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01211871
CAS-Nummer: 2585-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR