3-(4-Bromophenoxy)propanoic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Bromophenoxy)propanoic acid

OR962585-1G OR962585-5G
APOSOR962585-1GEA 115 EUR
APOSOR962585-1G APOSOR962585-5G
3-(4-Bromophenoxy)propanoic acid 98%
3-(4-Bromophenoxy)propanoic acid
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02295727
CAS-Nummer: 93670-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR