3-Pyrrolidinobenzyl alcohol 96%

Lieferant: Apollo Scientific

OR962686-1G OR962686-5G
APOSOR962686-1GEA 183 EUR
APOSOR962686-1G APOSOR962686-5G
3-Pyrrolidinobenzyl alcohol 96%
3-Pyrrolidinobenzyl alcohol
Formel: C₁₁H₁₅NO
Molecular Weight: 177.243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07772826
CAS-Nummer: 859850-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR