2-(1-Pyrrolidinyl)benzoic acid 97%
Lieferant: Apollo Scientific
Synonyme:
2-(Pyrrolidin-1-yl)benzoic acid
OR962734-1G
OR962734-5G
APOSOR962734-1GEA
77
EUR
APOSOR962734-1G
APOSOR962734-5G
2-(1-Pyrrolidinyl)benzoic acid 97%
2-(1-Pyrrolidinyl)benzoic acid
Formel:
C₁₁H₁₃NO₂ Molecular Weight: 191,226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09035985 CAS-Nummer: 78648-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...